missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Women's Curvy Fit Flame Resistant Jean with Stretch
Women's Curvy Fit Flame Resistant Jean with Stretch feel like an everyday jean. Light hand sanding and whiskering gives a more fashionable appearance. Felled inseam and seat seam adds durability. Contoured waistband provides a more comfortable fit.
Supplier: Bulwark™ PSJCSD2435U
Description
- Lightweight fabric with stretch that makes this work jean feel like an everyday jean.
- Light hand sanding and whiskering gives a more fashionable appearance.
- Felled inseam and seat seam adds durability.
- Contoured waistband provides a more comfortable fit.
- Metal button closure (metal is not exposed to the skin).
- Two patch on spade shaped hip pockets.
- Scoop front pockets with watch pocket on right side.
- Zipper fly closure.
- Certifications Level 1: NFPA 70E, NFPA 2112
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Arc Flash, Flash Fire
- Number of Pockets: 5
- Primary Closure: Concealed Button
- Wash Care: Industrial Laundry - Light Soil
- Fabric: Body: 12.5 oz. (425 g/m&superscript_2;) Flame Resistant Denim
- Blend: Body: 98% Cotton / 2% Spandex Elastane
- Waistband: Contoured waistband with stretch interlining
- Features: Contoured waistband, Stretch fabric, Hand sanded fabric with whiskering, Felled inseam and seat seam
- Protection: ATPV Arc Rating: 21
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Curvy
- Gender: Female
- Brand: Bulwark™
Specifications
24 | |
Cotton, Polyester, Spandex, Polyester, Spandex | |
Stitched | |
Concealed Button | |
Category 2 | |
NFPA 70E, NFPA 2112 |
Women's | |
Denim | |
4 | |
Reusable | |
21 | |
35 in. |