Learn More
DWK Life Sciences™ Wheaton™ Square HDPE Jug
Includes a 38-400 polyvinyl-lined screw cap.
$86.58 - $86.58
Specifications
Color Clear
Material HDPE
Quantity 4 Jugs/Cs.
Sterility Non-sterile
Graduated No
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
03-439-55B
View Documents
DWK Life Sciences™
222333
3785 mL
Case of 4 Each for $86.58
Only null left
Description
Description
  • Ideal for storage and transport of liquids
  • Integral handle supports carrying of the bottle
  • Good chemical resistance
Specifications
Specifications
Clear
4 Jugs/Cs.
No
Square
Polyvinyl
Screw Cap
HDPE
Non-sterile
Threaded
Square Jug
Polypropylene
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'