missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences™ Wheaton™ Serum Vial
Serum vials are converted from Type I borosilicate glass tubing.
Supplier: DWK Life Sciences™ W0UBB10MLAPEP
Description
- Vials conform to the hydrolytic resistance requirements outlined in USP 660 and EP 3.2.1
- Shrink-wrapped modules reduce particulate contamination
- Special design adds strength for freeze-drying applications
Specifications
Amber | |
Borosilicate Glass | |
Non-sterile | |
Autoclavable | |
150 Vials/Pk., 264 Pk./Cs. | |
No |
20 mm | |
No | |
10 mL | |
50 mm | |
Serum Vial |