missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Labeled Reagent Bottles
Autoclavable Reagent bottle manufactured from USP Type I borosilicate glass and features no-drip pour lip
$165.76 - $236.85
Specifications
Color | Clear |
---|---|
Includes | No-drip pour lip, large writing patch |
Material | Glass |
Mouth | Wide |
Product Type | Reagent Bottle |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
03-439-68A
|
DWK Life Sciences
220223 |
250 mL |
Case of 6 Each for $165.76
|
|
|||||
03-439-68B
|
DWK Life Sciences
220283 |
500 mL |
Case of 6 Each for $236.85
|
|
|||||
Description
- Pre-attached polypropylene screw cap with PTFE liner
- Large writing patch
Specifications
Clear | |
Glass | |
Reagent Bottle | |
Polypropylene |
No-drip pour lip, large writing patch | |
Wide | |
Polyethylene | |
Screw Cap |