Learn More
DWK Life Sciences Acura electro™ 956 8-Channel Pipetters - Basic Package
Large selection of 8-channel pipetters
$4505.35 - $4995.74
Specifications
Battery Type Rechargeable
Compatible Tips Universal fit pipette tip
Model 956
Number of Channels 8
Product Line Acura Electro
View More Specs
Includes: Acura electro basic package includes electronic pipette, individual QC certificate and operating instructions, pipette charging stand, fast charger with AC adapter and cord, additional battery pack, and pipette tip samples.
Products 4
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
03-410-012
View Documents
DWK Life Sciences
W870924A
0.5 to 10 μL
Case for $4,995.74
Only null left
03-410-013
View Documents
DWK Life Sciences
W870926A
2.5 to 50 μL
Case for $4,910.85
Only null left
03-410-015
View Documents
DWK Life Sciences
W870930A
20 to 350 μL
Case for $4,505.35
Only null left
03-410-014
View Documents
DWK Life Sciences
W870928A
10 to 200 μL
Case for $4,910.85
Only null left
Description
Description
360° rotation of multichannel volumetric module for best working position
  • Microprocessor-controlled motor
  • Rechargeable battery pack
  • Also fits any single channel volumetric module
  • Models cover the volume range 0.5 to 350μL
Specifications
Specifications
Rechargeable
956
Acura Electro
Digital
Electronic pipette, individual QC certificate and operating instructions, pipette charging stand, fast charger with AC adapter and cord, additional battery pack and pipette tip samples
Universal fit pipette tip
8
Pipetters Basic Package
Ergonomic
2 Years
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'