missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, USP Purified, Nonsterile, Ricca Chemical
$1433.50 - $1433.50
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | dihydrogen oxide, dihydrogen monoxide |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
36-101-5694
|
Ricca Chemical Company
R919100055E |
55 gal. | Poly Drum |
Each for $1,433.50
|
|
||||
Chemical Identifiers
7732-18-5 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
O |
H2O | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 |
Specifications
Colorless | |
98.0 | |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
100% | |
Poly Drum | |
Liquid |
7732-18-5 | |
100.0 | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
USP Purified | |
55 gal. | |
Water, Non-Sterile |