missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
7732-18-5 | |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
Plastic Bottle | |
1.00Â g/cm3 (at 20°C) |
100°C (1013Â hPa) | |
Deionized water | |
O | |
18.02 | |
23Â hPa (20°C) | |
1 L |
Chemical Identifiers
7732-18-5 | |
18.02 | |
Deionized water | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |
Safety and Handling
Recommended Storage : Store at 5 to 30°C