Learn More
Thermo Scientific Chemicals Water, Endotoxin-free
CAS: 7732-18-5 | H2O | 18.015 g/mol
$125.34 - $174.93
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.015 |
MDL Number | MFCD00011332 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | oxidane |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ65589AP
|
Thermo Scientific Chemicals
J65589AP |
500 mL |
Each for $125.34
|
|
|||||
AAJ65589K2
|
Thermo Scientific Chemicals
J65589K2 |
1 L |
Each for $174.93
|
|
|||||
Description
Water Endotoxin-free is used as solvent in industries and in all research fields. It is also used in laboratory for cell isolation and culture. It is also useful for elution of endotoxin- free plasmid DNA.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsWater Endotoxin-free is used as solvent in industries and in all research fields. It is also used in laboratory for cell isolation and culture. It is also useful for elution of endotoxin- free plasmid DNA.
Solubility
Miscible with methanol, acetone and ethanol.
Chemical Identifiers
7732-18-5 | |
18.015 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
H2O | |
MFCD00011332 | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
CHEBI:15377 | |
O |
Specifications
7 | |
32°F | |
H2O | |
2050024 | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02 | |
Odorless | |
500 mL | |
Liquid |
7732-18-5 | |
100°C | |
MFCD00011332 | |
1410039 | |
Miscible with methanol,acetone and ethanol. | |
O | |
18.015 | |
CHEBI:15377 | |
Endotoxin-Free | |
1 | |
1.34 | |
Water, Endotoxin-free |
Safety and Handling
EINECSNumber : 231-791-2
RTECSNumber : ZC0110000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only