missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, ACS Reagent Grade, ASTM Type II, Ricca Chemical
$32.07 - $932.53
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.015 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | dihydrogen oxide, dihydrogen monoxide |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | oxidane |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
R9152000500
|
Ricca Chemical Company
R9152000500A |
500 mL | Natural Poly Bottle |
Each for $32.07
|
|
||||
915232
|
Ricca Chemical Company
915232 |
1 L | Natural Poly Bottle |
Each for $63.22
|
|
||||
91521
|
Ricca Chemical Company
91521 |
4 L | Natural Poly Bottle |
Each for $70.81
|
|
||||
915225
|
Ricca Chemical Company
915225 |
10 L | Cubitainer |
Each for $131.38
|
|
||||
91525
|
Ricca Chemical Company
91525 |
20 L | Cubitainer |
Each for $188.83
|
|
||||
915255
|
Ricca Chemical Company
915255 |
55 gal. | Poly Drum |
Each for $932.53
|
|
||||
Chemical Identifiers
7732-18-5 | |
18.015 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
Specifications
7732-18-5 | |
100.0 | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
ACS | |
500 mL |
98.0 | |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
100% | |
Natural Poly Bottle | |
Water, ASTM Type II |