missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water with 0.1% Trifluoroacetic acid, LabReady™ Solvent Blend, for LC-MS and HPLC, Honeywell Burdick & Jackson
$734.68 - $867.25
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Synonym | water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
6002545
|
Honeywell-Burdick and Jackson
LC4851 |
6 x 1 L | Glass Bottle, Gl45 |
Case of 6 Each for $867.25
|
|
||||
6002544
|
Honeywell-Burdick and Jackson
LC48525 |
4 x 2.5 L | Glass Bottle, Gl45 |
Case of 4 Each for $734.68
|
|
||||
Chemical Identifiers
7732-18-5 | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide |
Specifications
7732-18-5,76-05-1 | |
99.9000%,0.1000% | |
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide | |
Glass Bottle, Gl45 | |
Water with 0.1% Trifluoroacetic acid, LabReady™ Solvent Blend, for LC-MS and HPLC |
100°C | |
0.9982 g/cm3 | |
HPLC | |
6 x 1 L |
Safety and Handling
ShelfLife : 365 days from date of manufacture