Learn More
Vikan™ Standard Broom Handle
The handle with attached item reaches to the chin of the user to prevent occupational back injuries.
Supplier: Vikan™ 29356
Description
- Made from aluminum and polypropylene
- Used with brooms, squeegees and scrapers
Specifications
Yellow | |
51 in. | |
Brooms, Squeegees and Scrapers | |
Yes |
Standard Broom Handle | |
Autoclavable | |
Hygienic (Euro) Thread | |
Aluminum, Polypropylene |