Learn More
Vikan™ Aluminum Handle
Used with all brooms, squeegees and scrapers.
Supplier:  Vikan™ 29373
Catalog No. 29-635-727
Only null left
Explore available promotions
Description
Description
  • Handle with the attached item should reach to the chin of the user to prevent occupational back injuries
Specifications
Specifications
Blue
Blue
Autoclavable
Yes
Handle
59.25 in.
1.25 in.
Aluminum
Product Suggestions
Product Suggestions