Learn More
MilliporeSigma™ Supelco™ VersaPak™ Silica Cartridges
Particle Size: 20 to 45 μm, Matrix Active Group: Spherical Silica
Supplier:  MilliporeSigma™ Supelco™ 97786U
 View more versions of this product
Catalog No. 111009100
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Spherical Silica
50 g
VersaPak™
Silica Cartridge
150 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
VersaPak is a registered trademark of Sigma-Aldrich Co. LLC