Learn More
Applied Biosystems™ Veriti™ Thermal Cycler + Plastics Package, 96-well, TriFlex plates
Applied Biosystems™ Veriti™ Thermal Cycler + Plastics Package with TriFlex Plates delivers the proven reliability of Applied Biosystems™ instruments with the throughput you need for large experiments.
Supplier: Applied Biosystems™ A41190
Description
- Convenient color touchscreen speeds setup time and use
- Transfer protocols, back up protocols and system configurations with a USB port
- From a single, networked Veriti™ instrument, start a run on up to 12 Veriti™ systems
- Conveniently place instruments side-by-side due to front to back venting feature
- Designed for use with the ProFlex™ 3 x 32-well PCR System and all Applied Biosystems standard 96-well thermal cyclers
- Easy-to-tear dual side-tabs for simple separation into three individual 32-well plates
- Semi-skirted for easy handling and labeling
- Standard 250 μL working volume, with a 300 μL maximum well volume when used with an adhesive film
- Alphanumeric labeling on each 32-well plate for easy well identification
- Certified human DNA-, DNase-, and RNase-free
Specifications
96-well Plate | |
Manual Protocols | |
up to 96 wells/run | |
Color Touchscreen | |
PCR | |
USB |
• Veriti 96-well Thermal Cycler • 5 x MicroAmp TriFlex 3 x 32-well PCR Reaction Plates (100 plates total) • 2 x MicroAmp 32-well Clear Adhesive Films (100 seals total) |
|
Fast, Standard | |
Applied Biosystems™ | |
Thermal Cycler | |
Approved for Shipment at Room Temperature or on Wet Ice |