missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Gold (Au) Standard for AAS 1000 ppm in 2% HCl, Ricca Chemical

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

$791.28 - $2258.53

Chemical Identifiers

CAS 16903-35-8
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RV010004100
View Documents
Ricca Chemical Company
RV010004100C
100 mL
Each for $791.28
Only null left
 
RV010004500
View Documents
Ricca Chemical Company
RV010004500C
500 mL
Each for $2,258.53
Only null left
 
Description

Description

  • 1000 ppm in 2% HCl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Chemical Identifiers

16903-35-8

Specifications

Amber Glass Bottle
95.87%,1.96%,0.17%
1.024
Approximately 0°C
High Purity
16903-35-8,7647-01-0,7732-18-5
99.79%,2.04%,0.17%
Liquid
100 mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'