missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ VeriShield™ Low Sound Management Dielectric Ear Muffs

VeriShield™ Low Sound Management Ear Muffs are available in over-the-head and hard hat-attached styles and enable the worker to hear voice and signal frequencies more naturally.

$20.62 - $20.62

Specifications

ANSI S3.1974 Yes
CSA Z94.2 Class B
Cushion Color Black
For Use With (Application) Construction, Electrical, Manufacturing, Oil and Gas, Mining, Steel and Metals, Agriculture
High Visibility No
View More Specs

Products 1
Catalog Number Mfr. No. Style Price Quantity  
Catalog Number Mfr. No. Style Price Quantity  
19-085-523
View Documents
Honeywell™
1035197VS
Over-the-head
Each for $20.62
Case of 10 Each for $60.10
Only null left
 
Description

Description

  • Enables workers to hear voices and signals more naturally
  • Available in Over-the-Head or Hard Hat-Attached styles
  • Durable, dialectric, non-deforming plastic headband
  • Memory foam ear cushions combine dense and soft foam
  • Reduces squeezing pressure on the head and enhances comfort
  • Durable construction
  • Oil-resistant cup surface withstands heavy-work environments and frequent cleaning
  • Designed for use with other PPE
Specifications

Specifications

Yes
Black
No
20 dB
Yes
Class B
Construction, Electrical, Manufacturing, Oil and Gas, Mining, Steel and Metals, Agriculture
1 Ea.
VeriShield
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'