missing translation for 'onlineSavingsMsg'
Learn More

Vanadium slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.8% (metals basis)

CAS: 7440-62-2 | V | 50.94 g/mol

$157.82 - $543.71

Chemical Identifiers

CAS 7440-62-2
Molecular Formula V
Molecular Weight (g/mol) 50.94
MDL Number MFCD00011453
InChI Key LEONUFNNVUYDNQ-UHFFFAOYSA-N
Synonym dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
PubChem CID 23990
ChEBI CHEBI:35170
IUPAC Name vanadium
SMILES [V]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4282806
View Documents
Thermo Scientific Chemicals
04282806
5 g
Each for $157.82
Only null left
 
AA4282814
View Documents
Thermo Scientific Chemicals
04282814
25 g
Each for $543.71
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-62-2
50.94
LEONUFNNVUYDNQ-UHFFFAOYSA-N
23990
vanadium
V
MFCD00011453
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
CHEBI:35170
[V]

Specifications

7440-62-2
Odorless
Slug
MFCD00011453
LEONUFNNVUYDNQ-UHFFFAOYSA-N
vanadium
23990
99.8%
(metals basis)
5 g
≈0.15g/slug
V
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
[V]
50.94
CHEBI:35170
Vanadium slug, 3.175mm (0.125 in.) dia. x 3.175mm (0.125 in.) length
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-171-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only