missing translation for 'onlineSavingsMsg'
Learn More

Valeric Acid, 99%, Spectrum™ Chemical

$303.97 - $1143.97

Chemical Identifiers

CAS 109-52-4
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18614635
View Documents
Spectrum Chemical Mfg Cor
VA110250ML
250 mL Amber Glass Bottle
Each for $303.97
Only null left
 
18614634
View Documents
Spectrum Chemical Mfg Cor
VA1101LT
1 L Amber Glass Bottle
Each for $493.50
Only null left
 
18614636
View Documents
Spectrum Chemical Mfg Cor
VA1104LT
4 L Amber Glass Bottle
Each for $1,143.97
Only null left
 
Description

Description

Specifications

Chemical Identifiers

109-52-4

Specifications

109-52-4
99%
250 mL
1
Reagent
Amber Glass Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.