missing translation for 'onlineSavingsMsg'
Learn More

Valco™ Precision Sampling Syringe, Magnum Series, Sold by MilliporeSigma™ Supelco™

Ideal for collection, trace analysis, and sample concentration of environmental contaminants

Supplier:  Merck Emd Millipore 21738

 View more versions of this product

Catalog No. 111003155


Only null left
Explore available promotions

Description

Description

  • Pressure-Lok series syringe allows sampling and storage of samples up to 500 mL
  • A low-dead volume, removable needle is used
  • Rotary valve allows on-site sampling for later analysis
Specifications

Specifications

Borosilicate Glass (Barrel)
0.023 in.
Collection, Trace Analysis and Sample Concentration of Environmental Contaminants
Precision Sampling Syringe
500 mL
Bevel
Magnum Series
0.035 in.
2 in.
1 Ea.
Valco
Removable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Valco is a registered trademark of Valco Instruments Co. Inc.