missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Vacufuge Plus - Centrifuge Concentrator PROMO
Facilitates fast and efficient, gentle vacuum concentration of DNA, RNA, nucleotides, proteins and other liquid or wet samples with its advanced heating technology. Its coated lid provides superior chemical resistance against many acids and organic solvents.
Supplier: Eppendorf™ 2231001136
Description
- Comes in 3 system configurations
- Extremely quiet operation <50 dB(A)
- Chemical-resistant, maintenance-free PTFE diaphragm pump eliminates the need for changing pump oil
- Small footprint saves valuable bench space
- Wide variety of 15 different rotor options to accommodate tubes from 0.2 mL to 50 mL, microplates and PCR plates
- Motorless induction drive and chemical-resistant, stainless steel chamber for trouble-free operation
- Can be connected with most commercially available cold traps for use with vapors
- Coated lid with superior chemical resistance
Specifications
Integrated Diaphragm Vacuum Pump | |
Vacufuge Plus | |
50/60 Hz | |
0.2 mL to 50 mL Tubes, Microplates and PCR Plates | |
PTFE, Stainless Steel |
Centrifuge Concentrator | |
120 V | |
Integrated Diaphragm Vacuum Pump | |
120 V, 50/60 Hz (US) |