missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Urine Collection System: Urine Tubes Only
Urine Tubes for use with T410 Urine Collection System.
Supplier: Fisherbrand™ T4101FS
Description
- Urine tubes are made of virgin polystyrene and are free of any mold release agents, metals or additives that could contaminate samples.
- They are made of heavy wall construction, graduated at 1/4, 1/2, 1, 2 and every 2 mL thereafter up to 12 mL, and can be safely centrifuged at speeds up to 2000 g.
- The tubes are flared at the top to make filling and drip-free pouring easier.
- Designed to allow the use of midget urinometers and reagent test strips requiring only 1/4 or 1/2 mL of sample.
- Size of tube: 105 mm H x 21 mm dia.
Specifications
Urine Collection Tube | |
Yes | |
Non-sterile | |
Urine Collection System, 36-101-5251 (T410) | |
4.1 in. | |
500 disposable 15 mL heavy-wall polystyrene tubes | |
500 | |
15 mL |
Single-use | |
Polystyrene | |
21 mm | |
0.25 to 12 mL | |
105 mm | |
Yes | |
0.5 oz. |