Learn More
Idex Peek Low-Pressure Union Assembly DFS Item
Make solid connections in low-pressure applications
Supplier:  Idex UPP702
 View more versions of this product
Catalog No. 29-008-654
Only null left
Explore available promotions
Description
Description
Make solid connections in low-pressure applications
  • Manufactured from inert polymers
  • 1/4-28 flat-bottom ports
  • Max pressure of 1000 psi (69 bar)
  • 0.030 in. thru-hole diameter
  • 0.41 μL swept volume
Specifications
Specifications
Union
2 P-200/P-235, 1/16 in., 1/4-28
PEEK
Thru Hole: 0.020 in.
1000 psi
80°C
Natural
Product Suggestions
Product Suggestions