Learn More
Idex Flat-Bottom Fittings: ETFE Ferrules
No need to flange tubing
Supplier: Idex UPP300NX
Description
Flangeless fitting eliminates the need to flange tubing. Just slide one of our Flangeless nuts onto your tubing with one of our Flangeless ferrules (coned end toward the nut) and finger tighten.
Specifications
0.19 in. | |
Standard Flat-Bottom Configurations | |
1/8 in. | |
Flangeless Ferrule |
0.19 in. | |
Natural | |
Tefzel |