missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Youngstown Barrel & Drum UltraTech Ultra-Pipe Sock™
Control the flow of sediment and oil
Supplier: Youngstown Barrel & Drum 9705OS
Description
Specifications
Gray | |
6.4 L | |
Pipe-sock | |
Sediment | |
1 Ea. | |
60 in. | |
8 in. | |
8 in. | |
60 in. | |
8 in. | |
2 lb. |
1.7 gal. | |
X-Tex | |
8 in. Dia. Pipe | |
Attached straps | |
60 x 8 x 8 in. | |
1624 mm | |
204 mm | |
204 mm | |
1624 mm | |
204 mm | |
1 kg |