missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biochrom Ultra Ninhydrin Reagent Kit
Biochrom Amino Acid Analysis reagents and buffers are manufactured from high quality chemicals and materials and subjected to rigorous control procedures, under the ISO 13485 Quality System.
$1310.87 - $5091.16
Specifications
Product Type | Reagent Kit |
---|---|
Content And Storage | Store at ambient temperature |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
BC80-2118-30
|
Biochrom
80211830 |
2 L |
Each for $1,310.87
|
|
|||||
BC80-2117-76
|
Biochrom
80211776 |
8 L |
Each for $5,091.16
|
|
|||||
Description
Biochrom buffers are guaranteed to give accurate and reproducible results for up to 3 years from manufacture, provided the storage conditions are adhered to. All reagents have stable chemistry and have been formulated to be stored at ambient temperature so, conveniently, do not need refrigeration.Specifications
Reagent Kit | |
Store at ambient temperature | |
Liquid |