missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Series 182B Coveralls, Bulk

Tyvek IsoClean delivers an ideal balance of protection, durability, and comfort. Bulk packaged in double transparent poly liners.

$342.37 - $415.45

Specifications

Product Type Cleanroom Coveralls
Material Flashspun Polyethylene
Color White
Disposable Single-use
Ankle Style Elastic
View More Specs

Products 7
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-088-1120E
View Documents
DuPont™
IC182BWH2X002500
2X-Large
Case of 25 Each for $362.90
Only null left
 
19-088-1120F
View Documents
DuPont™
IC182BWH3X002500
3X-Large
Case of 25 Each for $384.65
Only null left
 
19-088-1120G
View Documents
DuPont™
IC182BWH4X002500
4X-Large
Case of 25 Each for $415.45
Only null left
 
19-0881120C
View Documents
DuPont™
IC182BWHLG002500
Large
Case of 25 Each for $342.37
Only null left
 
19-088-1120B
View Documents
DuPont™
IC182BWHMD002500
Medium
Case of 25 Each for $342.37
Only null left
 
190881120A
View Documents
DuPont™
IC182BWHSM002500
Small
Case of 25 Each for $342.37
Only null left
 
19-0881120D
View Documents
DuPont™
IC182BWHXL002500
X-Large
Case of 25 Each for $342.37
Only null left
 
Description

Description

  • Made using a patented flash spinning process
  • Tyvek provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Elastic openings for tighter fit at wrist and ankle
  • Bound neck for lower particle shedding
  • Front zipper closure for easy donning and doffing
  • Raglan sleeve design for greater range of motion and comfort
Specifications

Specifications

Cleanroom Coveralls
White
Elastic
Zipper
Non-sterile
5 mil
Bulk
Flashspun Polyethylene
Single-use
Elastic
Bound
Raglan
Premium protection for controlled environments and inherent barrier to dry particles, microorganisms, and non-hazardous liquids
Class 1000 (ISO 6)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'