missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Series 108 Cleanroom Coveralls, Attached Hood and Boots
Cleanroom coveralls with attached hood and boots deliver an ideal balance of protection, durability, and comfort.
Supplier: DuPont™ IC108SWH5X0025TS
Description
- Unique, patented flash spinning process creates an inherent barrier to dry particles, microorganisms and non-hazardous liquids
- Comfortable, lightweight and durable
- Clean-processed garments offer lowest level of particle shedding within DuPont portfolio
- Sterile garment options are gamma sterilized to an SAL of 10-6
- Serged seam with covered elastic options
- IsoClean™ product portfolio
- Full traceability on all sterilized apparel
Specifications
Cleanroom Coveralls | |
Unisex | |
Single-use | |
Thumb Loop | |
Serged | |
Individually double bagged |
5X-Large | |
White | |
Elastic | |
Respirator Fit | |
Sterile | |
IC108SWH |