missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ 400 Comfort Fit Design 127S Series Coveralls, Vending Packaged
Coveralls with respirator fit hood provide breathable inherent barrier protection against hazardous dry particles, aerosols, and non-hazardous light liquid splash. Individually packaged for PPE vending machines.
$287.40 - $439.36
Specifications
Product Type | General Purpose Coveralls |
---|---|
Gender | Unisex |
Material | Tyvek |
Color | White |
Disposable | Limited-use |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-820-535
|
DuPont™
TY127SWHMD0025VP |
Medium |
Case of 25 Each for $287.40
|
|
|||||
19-820-534
|
DuPont™
TY127SWHLG0025VP |
Large |
Case of 25 Each for $287.40
|
|
|||||
19-820-536
|
DuPont™
TY127SWHXL0025VP |
X-Large |
Case of 25 Each for $287.40
|
|
|||||
19-820-528
|
DuPont™
TY127SWH2X0025VP |
2X-Large |
Case of 25 Each for $304.62
|
|
|||||
19-820-529
|
DuPont™
TY127SWH3X0025VP |
3X-Large |
Case of 25 Each for $322.91
|
|
|||||
19-820-530
|
DuPont™
TY127SWH4X0025VP |
4X-Large |
Case of 25 Each for $348.77
|
|
|||||
19-820-531
|
DuPont™
TY127SWH5X0025VP |
5X-Large |
Case of 25 Each for $376.66
|
|
|||||
19-820-532
|
DuPont™
TY127SWH6X0025VP |
6X-Large |
Case of 25 Each for $406.85
|
|
|||||
19-820-533
|
DuPont™
TY127SWH7X0025VP |
7X-Large |
Case of 25 Each for $439.36
|
|
|||||
Description
- Tyvek 400 coveralls provide an ideal balance of protection, durability and comfort of any limited use fabric technology.
- Fabric offers an inherent barrier against particles (down to 1.0 micron in size).
- Protection is built into the fabric itself; there are no films or laminates to abrade or wear away.
- Comfort fit design based on extensive wear.
- Enables a greater range of movement while stretching and bending, provides a more tailored fit.
- Longer zipper for easier donning and doffing and an elastic waist to better position the garment.
- Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
- Elastic opening for tighter fit at wrist, elastic opening for tighter fit at ankle
- Model 127 individually packaged for PPE vending machines
Specifications
General Purpose Coveralls | |
Tyvek | |
Limited-use | |
Elastic | |
Hooded | |
Front Zipper | |
Non-sterile | |
Yes | |
Lead and Asbestos Abatement, Remediation, General Maintenance, Operations, Spray Painting, Cleaning |
Unisex | |
White | |
Elastic | |
Elastic | |
Respirator Fit | |
Serged | |
ASTM D1777, D3776, D774, D5733, D5034, D257, AATCC 127 | |
TY127SWH | |
25/Cs. |