missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ 400 Comfort Fit Design 127S Series Coveralls, USDA, FDA, OSHA
Coveralls with respirator fit hood provide breathable inherent barrier protection against hazardous dry particles, aerosols, and non-hazardous light liquid splash. Meets NAFTA Standards. Meets USDA, FDA, OSHA Standards.
Supplier: DuPont™ TY127SWH4X0006G1
Description
- Tyvek 400 coveralls provide an ideal balance of protection, durability and comfort of any limited use fabric technology.
- Fabric offers an inherent barrier against particles (down to 1.0 micron in size).
- Protection is built into the fabric itself; there are no films or laminates to abrade or wear away.
- Comfort fit design based on extensive wear.
- Enables a greater range of movement while stretching and bending, provides a more tailored fit.
- Longer zipper for easier donning and doffing and an elastic waist to better position the garment.
- Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
- Elastic opening for tighter fit at wrist, elastic opening for tighter fit at ankle
- Model 127 USDA, FDA, OSHA compliant
Specifications
General Purpose Coveralls | |
Unisex | |
White | |
Elastic | |
Elastic | |
Respirator Fit | |
Serged | |
ASTM D1777, D3776, D774, D5733, D5034, D257, AATCC 127 | |
TY127SWH | |
25/Cs. |
4X-Large | |
Tyvek | |
Limited-use | |
Elastic | |
Hooded | |
Front Zipper | |
Non-sterile | |
Yes | |
Lead and Asbestos Abatement, Remediation, General Maintenance, Operations, Spray Painting, Cleaning |