Learn More
DuPont™ Tychem™ 6000 Coverall with Respirator Fit Hood, Sock Boots, Butyl Gloves
Tychem™ 6000 Coveralls provide protection from chemical warfare agents and toxic industrial chemicals.
$1357.56 - $1779.17
Specifications
Color Gray
Material Tychem
Ankle Style Boot Overflaps
Cuff Style Attached Gloves
Disposable Single-use
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
18-888-796
View Documents
DuPont™
TF199TGY4X0006WG
4X-Large
Case of 6 Each for $1,647.37
Only null left
18-888-797
View Documents
DuPont™
TF199TGY5X0006WG
5X-Large
Case of 6 Each for $1,779.17
Only null left
18-888-799
View Documents
DuPont™
TF199TGYMD0006WG
Medium
Case of 6 Each for $1,357.56
Only null left
18-888-800
View Documents
DuPont™
TF199TGYSM0006WG
Small
Case of 6 Each for $1,357.56
Only null left
18-888-794
View Documents
DuPont™
TF199TGY2X0006WG
2X-Large
Case of 6 Each for $1,439.02
Only null left
18-888-795
View Documents
DuPont™
TF199TGY3X0006WG
3X-Large
Case of 6 Each for $1,525.35
Only null left
18-888-798
View Documents
DuPont™
TF199TGYLG0006WG
Large
Case of 6 Each for $1,357.56
Only null left
18-888-801
View Documents
DuPont™
TF199TGYXL0006WG
X-Large
Case of 6 Each for $1,357.56
Only null left
Description
Description
Tychem™ 6000 coveralls are made of a proprietary barrier film laminated to a Tyvek™ substrate. Tychem™ 6000 fabrics are tested by a third party laboratory to assess its chemical permeation resistance.
  • Provides protection against over 180 chemical challenges for at least 30 minutes
  • Used by military personnel and first responders for chemical warfare situations
  • Available in low-visibility gray fabric for discretionary purpose
Specifications
Specifications
Gray
Boot Overflaps
Single-use
Yes
None
Liquid
Non-sterile
Tychem
Attached Gloves
Unisex
Hooded
6/Pk.
Taped
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'