Learn More
DuPont™ Tychem™ 5000 Bib Overall with Open Ankles
DuPont™ Tychem™ 5000 Bib Overall with Open Ankles provides protection against a broad range of chemicals and situations involving liquid splash.
$362.99 - $440.43
Specifications
Color | Tan |
---|---|
Seam Type | Taped |
Suspender | Adjustable Webbing Straps with Buckle Closure |
Ankle Style | Open |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
17-500-107
|
DuPont™
C3360TTN4X000600 |
4X-Large |
Case of 6 Each for $440.43
|
|
|||||
17-500-108
|
DuPont™
C3360TTNSM000600 |
Small |
Case of 6 Each for $362.99
|
|
|||||
Description
DuPont™ Tychem™ 5000 helps offer protection for those in hazardous environments.
- Provides at least 30 minutes of protection against over 150 chemical challenges
- NFPA 1992 certified
Specifications
Tan | |
Taped | |
Adjustable Webbing Straps with Buckle Closure | |
Open |