Learn More
DuPont™ Tychem™ 10000FR Encapsulated Level A Suit with Hansen Fitting
Tychem™ 10000 FR is a top choice for chemical protection in emergency situations.
$4,525.62 - $5,085.00
Specifications
Material Tychem
View More Specs
Products 3
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
18888757
View Documents
DuPont™
RF600TSV2X00017S
N/A
Only null left
18888760
View Documents
DuPont™
RF600TSV3X00017S
N/A
Only null left
18888770
View Documents
DuPont™
RF600TSVXL00017S
N/A
Only null left
Description
Description
Certified to NFPA 1991 in a single-skin construction, Tychem® 10000 FR garments are composed of a patented, heavy-duty aluminized fabric made of a DuPont™ Nomex™ and DuPont™ Kevlar™ blend, laminated to a multi-layer barrier film. Tychem® 10000 FR fabrics are tested by a third party laboratory to assess its chemical permeation resistance.
  • Provides at least 30 minutes of protection against over 285 chemical challenges
  • Ideally suited for industrial and HAZMAT situations
  • Tychem™ 10000 FR is high-visibility silver
  • A limited-use “single skin” garment offers broad chemical holdout
Certifications
NFPA 1991 with Flash Fire and Liquified Gas
Specifications
Specifications
Tychem
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'