missing translation for 'onlineSavingsMsg'
Learn More

Tungsten wire basket; # turns, 8; ID(mm), 7; Ht(mm), 9; Wire leads(cm), 5.0; Wire dia(mm), 0.51, Thermo Scientific™

$199.48 - $725.53

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA41178KK
View Documents
Thermo Scientific Chemicals
41178KK
10 Pc.
Each for $199.48
Only null left
 
AA41178KD
View Documents
Thermo Scientific Chemicals
41178KD
50 Pc.
Each for $725.53
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
23964
tungsten
W
MFCD00011461
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
CHEBI:27998
[W]

Specifications

7440-33-7
MFCD00011461
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
[W]
183.84
CHEBI:27998
Tungsten wire basket, 8 turns, ID(mm), 7; Ht(mm), 9; Wire leads(cm), 5.0; Wire dia(mm), 0.51
W
10 Pc.
WFKWXMTUELFFGS-UHFFFAOYSA-N
tungsten
23964
Wire
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-143-9

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'