missing translation for 'onlineSavingsMsg'
Learn More

Tris(dimethylamino)methane (stabilized with KOH) 97.0+%, TCI America™

$471.60 - $1616.14

Chemical Identifiers

CAS 5762-56-1
Molecular Formula C7H19N3
Molecular Weight (g/mol) 145.25
MDL Number MFCD00008322
InChI Key MUMVIYLVHVCYGI-UHFFFAOYSA-N
Synonym tris dimethylamino methane, n,n,n',n',n,n-hexamethylmethanetriamine, methanetriamine, n,n,n',n',n,n-hexamethyl, bis dimethylamino methyl dimethylamine, n,n,n,n,n,n-hexamethylmethanetriamine, trisdimethylaminomethane, acmc-209lzg, tris-dimethylaminomethane
PubChem CID 79831
IUPAC Name N,N,N',N',N'',N''-hexamethylmethanetriamine
SMILES CN(C)C(N(C)C)N(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T10575ML
View Documents
TCI America
T10575ML
5 mL
Each for $471.60
Only null left
 
T105725ML
View Documents
TCI America
T105725ML
25 mL
Each for $1,616.14
Only null left
 
Description

Description

Specifications

Chemical Identifiers

5762-56-1
145.25
MUMVIYLVHVCYGI-UHFFFAOYSA-N
79831
CN(C)C(N(C)C)N(C)C
C7H19N3
MFCD00008322
tris dimethylamino methane, n,n,n',n',n,n-hexamethylmethanetriamine, methanetriamine, n,n,n',n',n,n-hexamethyl, bis dimethylamino methyl dimethylamine, n,n,n,n,n,n-hexamethylmethanetriamine, trisdimethylaminomethane, acmc-209lzg, tris-dimethylaminomethane
N,N,N',N',N'',N''-hexamethylmethanetriamine

Specifications

5762-56-1
43°C
MFCD00008322
2733
MUMVIYLVHVCYGI-UHFFFAOYSA-N
N,N,N',N',N'',N''-hexamethylmethanetriamine
79831
≥97.0% (T)
Tris(dimethylamino)methane (stabilized with KOH)
Yellow
C7H19N3
5 mL
tris dimethylamino methane, n,n,n',n',n,n-hexamethylmethanetriamine, methanetriamine, n,n,n',n',n,n-hexamethyl, bis dimethylamino methyl dimethylamine, n,n,n,n,n,n-hexamethylmethanetriamine, trisdimethylaminomethane, acmc-209lzg, tris-dimethylaminomethane
CN(C)C(N(C)C)N(C)C
145.25
145.25
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'