missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(1H-benzotriazol-1-yl)methane 95.0+%, TCI America™
$123.03 - $438.05
Chemical Identifiers
CAS | 88088-95-3 |
---|---|
Molecular Formula | C19H13N9 |
Molecular Weight (g/mol) | 367.376 |
MDL Number | MFCD00514776 |
InChI Key | VENLCXAGJRTBFL-UHFFFAOYSA-N |
PubChem CID | 3733734 |
IUPAC Name | 1-[bis(benzotriazol-1-yl)methyl]benzotriazole |
SMILES | C1=CC=C2C(=C1)N=NN2C(N3C4=CC=CC=C4N=N3)N5C6=CC=CC=C6N=N5 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
T3141200MG
|
TCI America
T3141200MG |
200 mg |
Each for $123.03
|
|
|||||
T31411G
|
TCI America
T31411G |
1 g |
Each for $438.05
|
|
|||||
Chemical Identifiers
88088-95-3 | |
367.376 | |
VENLCXAGJRTBFL-UHFFFAOYSA-N | |
1-[bis(benzotriazol-1-yl)methyl]benzotriazole |
C19H13N9 | |
MFCD00514776 | |
3733734 | |
C1=CC=C2C(=C1)N=NN2C(N3C4=CC=CC=C4N=N3)N5C6=CC=CC=C6N=N5 |
Specifications
88088-95-3 | |
White-Yellow | |
MFCD00514776 | |
VENLCXAGJRTBFL-UHFFFAOYSA-N | |
1-[bis(benzotriazol-1-yl)methyl]benzotriazole | |
3733734 | |
≥95.0% (HPLC) | |
Tris(1H-benzotriazol-1-yl)methane |
191°C | |
C19H13N9 | |
200 mg | |
C1=CC=C2C(=C1)N=NN2C(N3C4=CC=CC=C4N=N3)N5C6=CC=CC=C6N=N5 | |
367.376 | |
367.38 | |
Crystalline Powder |
Safety and Handling
TSCA : No