missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrippNT LabBox Organizers
$316.27 - $460.57
Specifications
For Use With (Application) | For samples, drop-offs and safety glass storage |
---|---|
Size | Small |
Product Type | Lab Box Organizer |
Catalog Number | Mfr. No. | Style | Dimensions (D x W x H) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Style | Dimensions (D x W x H) | Price | Quantity | ||||
50856163
![]() |
TrippNT
TRP50081 |
Double Facing Tape Backing | 6 x 9 x 6 in. |
Each for $316.27
|
|
||||
50856162
![]() |
TrippNT
TRP50116 |
Magnetic Backing | 6 x 9 x 6 in. |
Each for $405.23
|
|
||||
50856158
![]() |
TrippNT
TRP50358 |
Magnetic Backing | 6 x 9 x 9 in. |
Each for $460.57
|
|
||||
Description
Lab boxes for samples, drop-offs and safety glass storage. Wall-mountable. Keyholes provided for permanent installation.
Specifications
For samples, drop-offs and safety glass storage | |
Small | |
Lab Box Organizer |