missing translation for 'onlineSavingsMsg'
Learn More

Trimethylphenylammonium Bromide 98.0+%, TCI America™

$71.81 - $487.56

Chemical Identifiers

CAS 16056-11-4
Molecular Formula C9H14BrN
Molecular Weight (g/mol) 216.122
MDL Number MFCD00011788
InChI Key GNMJFQWRASXXMS-UHFFFAOYSA-M
Synonym trimethylphenylammonium bromide, phenyltrimethylammonium bromide, n,n,n-trimethylbenzenaminium bromide, benzenaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethylanilinium bromide, phenyl trimethyl ammonium bromide, ammonium, phenyltrimethyl-, bromide, trimethyl phenyl azanium bromide, benzenaminium, n,n,n-trimethyl-, bromide 1:1, ammonium trimethylphenyl-, bromide
PubChem CID 27663
IUPAC Name trimethyl(phenyl)azanium;bromide
SMILES C[N+](C)(C)C1=CC=CC=C1.[Br-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P024325G
View Documents
TCI America
P024325G
25 g
Each for $71.81
Only null left
 
P0243100G
View Documents
TCI America
P0243100G
100 g
Each for $164.41
Only null left
 
P0243500G
View Documents
TCI America
P0243500G
500 g
Each for $487.56
Only null left
 
Description

Description

Specifications

Chemical Identifiers

16056-11-4
216.122
GNMJFQWRASXXMS-UHFFFAOYSA-M
27663
C[N+](C)(C)C1=CC=CC=C1.[Br-]
C9H14BrN
MFCD00011788
trimethylphenylammonium bromide, phenyltrimethylammonium bromide, n,n,n-trimethylbenzenaminium bromide, benzenaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethylanilinium bromide, phenyl trimethyl ammonium bromide, ammonium, phenyltrimethyl-, bromide, trimethyl phenyl azanium bromide, benzenaminium, n,n,n-trimethyl-, bromide 1:1, ammonium trimethylphenyl-, bromide
trimethyl(phenyl)azanium;bromide

Specifications

16056-11-4
C9H14BrN
25 g
GNMJFQWRASXXMS-UHFFFAOYSA-M
trimethyl(phenyl)azanium;bromide
27663
≥98.0% (HPLC,T)
Trimethylphenylammonium Bromide
White
MFCD00011788
trimethylphenylammonium bromide, phenyltrimethylammonium bromide, n,n,n-trimethylbenzenaminium bromide, benzenaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethylanilinium bromide, phenyl trimethyl ammonium bromide, ammonium, phenyltrimethyl-, bromide, trimethyl phenyl azanium bromide, benzenaminium, n,n,n-trimethyl-, bromide 1:1, ammonium trimethylphenyl-, bromide
C[N+](C)(C)C1=CC=CC=C1.[Br-]
216.122
216.12
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1187

RTECSNumber : BT2100000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'