missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylene Sulfide 98.0+%, TCI America™
$251.65 - $836.97
Chemical Identifiers
CAS | 287-27-4 |
---|---|
Molecular Formula | C3H6S |
Molecular Weight (g/mol) | 74.14 |
MDL Number | MFCD00042751 |
InChI Key | XSROQCDVUIHRSI-UHFFFAOYSA-N |
Synonym | trimethylene sulfide, thiacyclobutane, propane, 1,3-epithio, unii-1o7c19qqdb, 1o7c19qqdb, 1,3-epithio propane, acmc-209h4c, trimethylene sulfide 8ci, trimethylene sulfide gc, inchi=1/c3h6s/c1-2-4-3-1/h1-3h |
PubChem CID | 9251 |
ChEBI | CHEBI:87565 |
IUPAC Name | thietane |
SMILES | C1CSC1 |
Chemical Identifiers
287-27-4 | |
74.14 | |
XSROQCDVUIHRSI-UHFFFAOYSA-N | |
9251 | |
thietane |
C3H6S | |
MFCD00042751 | |
trimethylene sulfide, thiacyclobutane, propane, 1,3-epithio, unii-1o7c19qqdb, 1o7c19qqdb, 1,3-epithio propane, acmc-209h4c, trimethylene sulfide 8ci, trimethylene sulfide gc, inchi=1/c3h6s/c1-2-4-3-1/h1-3h | |
CHEBI:87565 | |
C1CSC1 |
Specifications
287-27-4 | |
94°C | |
MFCD00042751 | |
1993 | |
XSROQCDVUIHRSI-UHFFFAOYSA-N | |
thietane | |
9251 | |
74.14 | |
Liquid |
Yellow | |
C3H6S | |
5 mL | |
trimethylene sulfide, thiacyclobutane, propane, 1,3-epithio, unii-1o7c19qqdb, 1o7c19qqdb, 1,3-epithio propane, acmc-209h4c, trimethylene sulfide 8ci, trimethylene sulfide gc, inchi=1/c3h6s/c1-2-4-3-1/h1-3h | |
C1CSC1 | |
74.14 | |
CHEBI:87565 | |
≥98.0% (GC) | |
Trimethylene Sulfide |
Safety and Handling
TSCA : Yes