missing translation for 'onlineSavingsMsg'
Learn More

Trimethoprim, 50 mg/ml in DMSO, sterile-filtered, Thermo Scientific Chemicals

$282.45 - $953.26

Chemical Identifiers

CAS 738-70-5
Molecular Formula C14H18N4O3
Molecular Weight (g/mol) 290.323
MDL Number MFCD00036761
InChI Key IEDVJHCEMCRBQM-UHFFFAOYSA-N
PubChem CID 5578
ChEBI CHEBI:45924
IUPAC Name 5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
SMILES COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67175XF
View Documents
Thermo Scientific Chemicals
J67175XF
1 mL
Each for $282.45
Only null left
 
AAJ671758EQ
View Documents
Thermo Scientific Chemicals
J671758EQ
5 x 1 mL
Pack for $953.26
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

738-70-5
290.323
IEDVJHCEMCRBQM-UHFFFAOYSA-N
CHEBI:45924
COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N
C14H18N4O3
MFCD00036761
5578
5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine

Specifications

Trimethoprim
738-70-5
1 mL
MFCD00036761
COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N
290.323
CHEBI:45924
Liquid
Plastic Bottle
C14H18N4O3
IEDVJHCEMCRBQM-UHFFFAOYSA-N
5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
5578
Trimethoprim
Videos
Safety and Handling

Safety and Handling

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 212-006-2

RTECSNumber : UV8225000

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only