missing translation for 'onlineSavingsMsg'
Learn More

TRIM10 Monoclonal Antibody (K92040_4A6), Invitrogen™

Mouse Monoclonal Antibody

$1421.98 - $2134.69

Specifications

Antigen TRIM10
Clone K92040_4A6
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MA556015
View Documents
Invitrogen™
MA556015
200 μg
Each for $2,134.69
Only null left
 
MA555635
View Documents
Invitrogen™
MA555635
50 μg
Each for $1,421.98
Only null left
 
Description

Description

Sequence of this protein is as follows: LEESPTCPLC KEPFRPGSFR PNWQLANVVE NIERLQLVST LGLGEEDVCQ EHGEKIYFFC EDDEMQLCVV CREAGEHATH TMRFLEDAAA PYREQIHKCL KCLRKEREEI QEIQSRENKR

TRIM10 Monoclonal antibody specifically detects TRIM10 in Human, Rat samples. It is validated for Western Blot
Specifications

Specifications

TRIM10
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide
AI324236; B30-RING finger protein; BB139825; Hematopoietic RING finger 1; Herf1; RFB30; RING finger protein 9; Rnf9; TRIM10; tripartite motif containing 10; tripartite motif protein 10; tripartite motif-containing 10; tripartite motif-containing protein 10; Zn-finger protein
Recombinant human TRIM10 (immunogen range is 51-170).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
TRIM10
K92040_4A6
Western Blot
Unconjugated
Mouse
Human, Rat
Q9UDY6
10107, 294210
IgG1
Protein A
Antibody
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'