missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trihexyl Borate 98.0+%, TCI America™
Supplier: TCI America B209425G
Specifications
Trihexyl Borate | |
Yellow | |
C18H39BO3 | |
25 g | |
KDQYHGMMZKMQAA-UHFFFAOYSA-N | |
trihexyl borate | |
21423 | |
≥98.0% (T) |
5337-36-0 | |
311°C | |
MFCD00027295 | |
Boric Acid Trihexyl Ester | |
B(OCCCCCC)(OCCCCCC)OCCCCCC | |
314.317 | |
314.32 | |
Liquid |
Chemical Identifiers
5337-36-0 | |
314.317 | |
KDQYHGMMZKMQAA-UHFFFAOYSA-N | |
21423 | |
B(OCCCCCC)(OCCCCCC)OCCCCCC |
C18H39BO3 | |
MFCD00027295 | |
Boric Acid Trihexyl Ester | |
trihexyl borate |
Safety and Handling
RTECSNumber : ED5120000
TSCA : Yes