missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic Acid, Reagents

Trifluoroacetic Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$584.53 - $8279.50

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym Perfluoroacetic Acid, Trifluoroethanoic Acid
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-336-908
View Documents
Labchem Inc
C234266025Y
2.5 L
Each for $8,279.50
Only null left
 
01-336-907
View Documents
Labchem Inc
C2342660500X
500 mL
Each for $584.53
Only null left
 
Description

Description

Specifications

Chemical Identifiers

76-05-1
114.02
DTQVDTLACAAQTR-UHFFFAOYSA-N
trifluoroacetic acid
C2HF3O2
MFCD00004169
Perfluoroacetic Acid, Trifluoroethanoic Acid
OC(=O)C(F)(F)F

Specifications

<1
76-05-1
Colorless to Slight Yellow
MFCD00004169
DTQVDTLACAAQTR-UHFFFAOYSA-N
trifluoroacetic acid
Reagent
Glass Bottle
1.535
-15.4°C
72.4°C
C2HF3O2
Perfluoroacetic Acid, Trifluoroethanoic Acid
OC(=O)C(F)(F)F
114.02
2.5 L
Liquid
Trifluoroacetic Acid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'