Learn More
Triethylene glycol dimethacrylate, contains MEHQ as inhibitor
CAS: 109-16-0 | C14H22O6 | 286.32 g/mol
$140.33 - $448.19
Chemical Identifiers
CAS | 109-16-0 |
---|---|
Molecular Formula | C14H22O6 |
Molecular Weight (g/mol) | 286.32 |
InChI Key | HWSSEYVMGDIFMH-UHFFFAOYSA-N |
IUPAC Name | 2-(2-{2-[(2-methylprop-2-enoyl)oxy]ethoxy}ethoxy)ethyl 2-methylprop-2-enoate |
SMILES | CC(=C)C(=O)OCCOCCOCCOC(=O)C(C)=C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC466272500
|
Thermo Scientific Chemicals
466272500 |
250 mL |
Each for $140.33
|
|
|||||
AC466270010
|
Thermo Scientific Chemicals
466270010 |
1 L |
Each for $448.19
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
109-16-0 | |
286.32 | |
2-(2-{2-[(2-methylprop-2-enoyl)oxy]ethoxy}ethoxy)ethyl 2-methylprop-2-enoate |
C14H22O6 | |
HWSSEYVMGDIFMH-UHFFFAOYSA-N | |
CC(=C)C(=O)OCCOCCOCCOC(=O)C(C)=C |
Specifications
109-16-0 | |
170.0°C to 172.0°C (5.0 mmHg) | |
C14H22O6 | |
1.092 | |
HWSSEYVMGDIFMH-UHFFFAOYSA-N | |
2-(2-{2-[(2-methylprop-2-enoyl)oxy]ethoxy}ethoxy)ethyl 2-methylprop-2-enoate | |
11 mPa.s (25°C) | |
Triethylene glycol dimethacrylate |
1.0920 g/mL | |
167°C | |
250 mL | |
Solubility aq. soln.: 3.6 g/L (20°C). Other solubilities: miscible with most organic solvents | |
CC(=C)C(=O)OCCOCCOCCOC(=O)C(C)=C | |
286.32 | |
286.32 |
Safety and Handling
GHS H Statement May cause an allergic skin reaction.
GHS P Statement Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 203-652-6
RUO – Research Use Only