missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Fluka™ Triethylammonium Bicarbonate Buffer, Honeywell™ Fluka™
volatile buffer, for HPLC, 1Â M
$422.68 - $422.68
Specifications
Color | Colorless |
---|---|
For Use With (Equipment) | Volatile buffer for HPLC |
Molecular Weight (g/mol) | 163.217 |
pH | 8.4 to 8.6 (at 25°C) |
Packaging | Glass Bottle |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Colorless | |
163.217 | |
Glass Bottle | |
Liquid |
Volatile buffer for HPLC | |
8.4 to 8.6 (at 25°C) | |
15715-58-9 |