missing translation for 'onlineSavingsMsg'
Learn More

Trichloroethylene, ACS, 99.5%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor T1115200LTBL

 View more versions of this product

Catalog No. 18614526

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

79-01-6
Drum
200 L
XSTXAVWGXDQKEL-UHFFFAOYSA-N
1,1,2-trichloroethene
0.995
0.0001 meq/g
1
C2HCl3
1e-05
ClC=C(Cl)Cl
131.38
ACS

Chemical Identifiers

79-01-6
131.38
1,1,2-trichloroethene
C2HCl3
XSTXAVWGXDQKEL-UHFFFAOYSA-N
ClC=C(Cl)Cl
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.