Learn More
Trichloroethylene, 99.6%, ACS reagent, stabilized
CAS: 79-01-6 | C2HCl3 | 131.39 g/mol
$437.54 - $437.54
Chemical Identifiers
CAS | 79-01-6 |
---|---|
Molecular Formula | C2HCl3 |
Molecular Weight (g/mol) | 131.39 |
MDL Number | MFCD00000838 |
InChI Key | XSTXAVWGXDQKEL-UHFFFAOYSA-N |
Synonym | trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen |
PubChem CID | 6575 |
ChEBI | CHEBI:16602 |
IUPAC Name | 1,1,2-trichloroethene |
SMILES | C(=C(Cl)Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC421520025
|
Thermo Scientific Chemicals
421520025 |
2.5 L | Glass bottle |
Each for $437.54
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
79-01-6 | |
131.39 | |
XSTXAVWGXDQKEL-UHFFFAOYSA-N | |
6575 | |
1,1,2-trichloroethene |
C2HCl3 | |
MFCD00000838 | |
trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen | |
CHEBI:16602 | |
C(=C(Cl)Cl)Cl |
Specifications
79-01-6 | |
1.4600g/mL | |
99.5% min. (GC) | |
C2HCl3 | |
ClCH=CCl2 | |
2.5 L | |
0.001% max. | |
1.46 | |
trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen | |
XSTXAVWGXDQKEL-UHFFFAOYSA-N | |
1,1,2-trichloroethene | |
0.5% (typically) Ethanol, or 0.8% (typically) Ethyloxirane | |
0.55 mPa.s (25°C) | |
131.39 | |
ACS Reagent | |
Liquid |
-85°C | |
87.0°C | |
Glass bottle | |
1.4760 to 1.4780 | |
MFCD00000838 | |
01, 187 | |
09,479; 10,414; 11,552 | |
15, 9804 | |
Solubility in water: insoluble. Other solubilities: miscible with most common organic solvents | |
C(=C(Cl)Cl)Cl | |
131.39 | |
6575 | |
CHEBI:16602 | |
99.6% | |
0.0001 meq/g max | |
Trichloroethylene |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness.
Harmful to aquatic life with long lasting effects.
May cause cancer.
Suspected of causing genetic defects.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at r
GHS Signal Word: Danger
EINECSNumber : 201-167-4
RTECSNumber : KX4550000
TSCA : TSCA
RUO – Research Use Only