missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroethylene, ≥99.5%, ACS Reagent, Honeywell™ Riedel-de Haën™
ACS Reagent,≥99.5%
$287.83 - $287.83
Chemical Identifiers
CAS | 79-01-6 |
---|---|
Molecular Formula | C2HCl3 |
Molecular Weight (g/mol) | 131.38 |
MDL Number | MFCD00000838 |
InChI Key | XSTXAVWGXDQKEL-UHFFFAOYSA-N |
Synonym | trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen |
PubChem CID | 6575 |
ChEBI | CHEBI:16602 |
IUPAC Name | 1,1,2-trichloroethene |
SMILES | C(=C(Cl)Cl)Cl |
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
79-01-6 | |
131.38 | |
XSTXAVWGXDQKEL-UHFFFAOYSA-N | |
6575 | |
1,1,2-trichloroethene |
C2HCl3 | |
MFCD00000838 | |
trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen | |
CHEBI:16602 | |
C(=C(Cl)Cl)Cl |
Specifications
79-01-6 | |
1.463g/mL at 25°C | |
770°F | |
C2HCl3 | |
MFCD00000838 | |
UN1710 | |
trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen | |
C(=C(Cl)Cl)Cl | |
131.38 | |
6575 | |
131.39g/mol | |
≥99.5% | |
Liquid |
-84.8°C | |
86.7°C | |
Glass Bottle | |
ClCH=CCl2 | |
1 L | |
1736782 | |
XSTXAVWGXDQKEL-UHFFFAOYSA-N | |
1,1,2-trichloroethene | |
4.5 (vs air) | |
CHEBI:16602 | |
61mmHg (20°C) | |
ACS Reagent | |
Trichloroethylene |
Safety and Handling
P201-P261-P273-P281-P305 + P351 + P338-P308 + P313
H315-H319-H336-H341-H350-H412
DOTInformation : 6.1/PGIII
EINECSNumber : 201-167-4
RTECSNumber : KX4550000