missing translation for 'onlineSavingsMsg'
Learn More

Trichloroacetic acid, 5% w/v aq. soln.

Protein precipitation reagent | CAS: 76-03-9 | C2HCl3O2 | 163.378 g/mol

$142.45 - $196.86

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.378
MDL Number MFCD00004177
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62366AP
View Documents
Thermo Scientific Chemicals
J62366AP
500 mL N/A
Only null left
 
AAJ62366K2
View Documents
Thermo Scientific Chemicals
J62366K2
1 L N/A
Only null left
 
Description

Description

Trichloroacetic acid, is traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. It is also used as a decalcifier and fixative in microscopy in protein sequencing, detecting albumin and organic synthesis. A protocol for the precipitation of nucleic acids can be found in Molecular Cloning.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trichloroacetic acid, is traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. It is also used as a decalcifier and fixative in microscopy in protein sequencing, detecting albumin and organic synthesis. A protocol for the precipitation of nucleic acids can be found in Molecular Cloning.

Solubility
Soluble in water (1200 mg/ml at 20°C), alcohol, ether, ethanol (very soluble), and carbon tetrachloride.

Notes
Store under nitrogen. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong bases, smines.
Specifications

Chemical Identifiers

76-03-9
163.378
YNJBWRMUSHSURL-UHFFFAOYSA-N
6421
2,2,2-trichloroacetic acid
C2HCl3O2
MFCD00004177
trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
CHEBI:30956
C(=O)(C(Cl)(Cl)Cl)O

Specifications

76-03-9
C2HCl3O2
MFCD00004177
970119
trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
YNJBWRMUSHSURL-UHFFFAOYSA-N
2,2,2-trichloroacetic acid
163.378
CHEBI:30956
500 mL
Liquid
Colorless
Cl3CCO2H
UN2564
14,9627
Soluble in water (1200mg/ml at 20°C),alcohol,ether,ethanol (very soluble),and carbon tetrachloride.
C(=O)(C(Cl)(Cl)Cl)O
5% w/v aq. soln.
6421
163.39
Acrid
Trichloroacetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335-H351

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: TRICHLOROACETIC ACID, SOLUTION

EINECSNumber : 200-927-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only