missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributyl Borate 99.0+%, TCI America™
$41.02 - $212.51
Chemical Identifiers
CAS | 688-74-4 |
---|---|
Molecular Formula | C12H27BO3 |
Molecular Weight (g/mol) | 230.155 |
MDL Number | MFCD00009434 |
InChI Key | LGQXXHMEBUOXRP-UHFFFAOYSA-N |
Synonym | n-butyl borate, tri-n-butyl borate, tributoxyborane, butyl borate, tributoxyboron, boron tributoxide, borester 2, tri-n-butoxyborane, tris butoxy borane, borane, tributoxy |
PubChem CID | 12712 |
IUPAC Name | tributyl borate |
SMILES | B(OCCCC)(OCCCC)OCCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B051825ML
|
TCI America
B051825ML |
25 mL |
Each for $41.02
|
|
|||||
B0518100ML
|
TCI America
B0518100ML |
100 mL |
Each for $80.16
|
|
|||||
B0518500ML
|
TCI America
B0518500ML |
500 mL |
Each for $212.51
|
|
|||||
Chemical Identifiers
688-74-4 | |
230.155 | |
LGQXXHMEBUOXRP-UHFFFAOYSA-N | |
12712 | |
B(OCCCC)(OCCCC)OCCCC |
C12H27BO3 | |
MFCD00009434 | |
n-butyl borate, tri-n-butyl borate, tributoxyborane, butyl borate, tributoxyboron, boron tributoxide, borester 2, tri-n-butoxyborane, tris butoxy borane, borane, tributoxy | |
tributyl borate |
Specifications
688-74-4 | |
230°C | |
MFCD00009434 | |
n-butyl borate, tri-n-butyl borate, tributoxyborane, butyl borate, tributoxyboron, boron tributoxide, borester 2, tri-n-butoxyborane, tris butoxy borane, borane, tributoxy | |
B(OCCCC)(OCCCC)OCCCC | |
230.155 | |
230.16 | |
Liquid |
Colorless | |
C12H27BO3 | |
25 mL | |
LGQXXHMEBUOXRP-UHFFFAOYSA-N | |
tributyl borate | |
12712 | |
≥99.0% (T) | |
Tributyl Borate |
Safety and Handling
EINECSNumber : (2)-2101
RTECSNumber : ED4900000
TSCA : Yes