missing translation for 'onlineSavingsMsg'
Learn More

trans-Cinnamaldehyde 98.0+%, TCI America™

$29.83 - $78.29

Chemical Identifiers

CAS 14371-10-9
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00007000
InChI Key KJPRLNWUNMBNBZ-QPJJXVBHSA-N
Synonym cinnamaldehyde, trans-cinnamaldehyde, cinnamic aldehyde, e-cinnamaldehyde, cinnamal, 3-phenylacrylaldehyde, zimtaldehyde, cinnamylaldehyde, 3-phenylpropenal, 2e-3-phenylprop-2-enal
PubChem CID 637511
ChEBI CHEBI:16731
IUPAC Name (2E)-3-phenylprop-2-enal
SMILES O=C\C=C\C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C035225ML
View Documents
TCI America
C035225ML
25 mL
Each for $29.83
Only null left
 
C0352500ML
View Documents
TCI America
C0352500ML
500 mL
Each for $78.29
Only null left
 
Description

Description

Specifications

Chemical Identifiers

14371-10-9
132.16
KJPRLNWUNMBNBZ-QPJJXVBHSA-N
637511
(2E)-3-phenylprop-2-enal
C9H8O
MFCD00007000
cinnamaldehyde, trans-cinnamaldehyde, cinnamic aldehyde, e-cinnamaldehyde, cinnamal, 3-phenylacrylaldehyde, zimtaldehyde, cinnamylaldehyde, 3-phenylpropenal, 2e-3-phenylprop-2-enal
CHEBI:16731
O=C\C=C\C1=CC=CC=C1

Specifications

14371-10-9
Yellow
C9H8O
25 mL
KJPRLNWUNMBNBZ-QPJJXVBHSA-N
(2E)-3-phenylprop-2-enal
637511
132.16
Liquid
-8°C
252°C
MFCD00007000
cinnamaldehyde, trans-cinnamaldehyde, cinnamic aldehyde, e-cinnamaldehyde, cinnamal, 3-phenylacrylaldehyde, zimtaldehyde, cinnamylaldehyde, 3-phenylpropenal, 2e-3-phenylprop-2-enal
O=C\C=C\C1=CC=CC=C1
132.16
CHEBI:16731
≥98.0% (GC)
trans-Cinnamaldehyde
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1148

RTECSNumber : GD6476000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'